To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:ALX Homeobox 1
- Antigen Symbol:ALX1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Gene ID:8092
- Antigen Synonyms:HEL23|CART1|FND3
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10279-084
- Supplier no.:BS-12356R-A350
Specifications
About this item
ALX1 is a 326 amino acid protein specific to cervix and cartilage tissues. As well as having a homeobox domain, ALX1 also contains an OAR domain, which has been suggested to be important for DNA binding or protein-protein interactions and transactivation. First characterized from a rat chondrosarcoma tumor cell line, ALX1 is a homeobox transcription factor that regulates downstream target genes and has specifically shown to act as a transcriptional repressor for rat prolactin in vivo. Homozygous ALX1 deficient mice are born with acrania and meroanencephaly, suggesting ALX1 function in the development of the neural tube. It has also been suggested that ALX1 cooperates with ALX4 in limb development and craniofacial bone formation.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ALX1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat