Order Entry
United States
ContactUsLinkComponent
Anti-TNF-related Apoptosis-inducing Ligand Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anti-TNF-related Apoptosis-inducing Ligand Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # 10274-722
Supplier:  Avantor
Anti-TNF-related Apoptosis-inducing Ligand Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # 10274-722
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Tumor Necrosis Factor (Ligand) Superfamily, Member 10
  • Antigen Symbol:
    TNFSF10
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 488
  • Flow Cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Size:
    100 µL
  • Environmentally Preferable:
  • Cross Adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    8743
  • Antigen Synonyms:
    Tumor necrosis factor ligand superfamily member 10|Apo-2 ligand|Apo-2L|Protein TRAIL|TNF-related apoptosis-inducing ligand|CD253
  • Storage Buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage Temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping Temperature:
    4°C
  • Immunogen:
    215-265/281
  • Purification:
    Purified by Protein A
  • Cat. No.:
    10274-722
  • Supplier no.:
    BS-1214R-A488

Specifications

About this item

Cytokine that binds to TNFRSF10A/TRAILR1, TNFRSF10B/TRAILR2, TNFRSF10C/TRAILR3, TNFRSF10D/TRAILR4 and possibly also to TNFRSF11B/OPG. Induces apoptosis. Its activity may be modulated by binding to the decoy receptors TNFRSF10C/TRAILR3, TNFRSF10D/TRAILR4 and TNFRSF11B/OPG that cannot induce apoptosis.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: TNF-related Apoptosis-inducing Ligand
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat