To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Shootin 1
- Antigen Symbol:SHTN1
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Antigen Synonyms:Putative uncharacterized protein KIAA1598|DKFZp686A0439|SHOT1_HUMAN.|Shootin 1|MGC40476|hypothetical protein FLJ11122|Shootin-1
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10264-790
- Supplier no.:BS-11863R-CY5
Specifications
About this item
Shootin1 is a 631 amino acid protein that belongs to the shootin family. The shootin1 protein contains three coiled-coil domains, a proline-rich region and interacts with RUFY3. Shootin1 is involved in the generation of internal asymmetric signals required for neuronal polarization. The shootin1 protein acts upstream of PI3K (phosphoinositide 3-kinase), by being required for spatially localized PI3K activity. By accumulating asymmetrically in a single neurite before polarization, shootin1 leads to axon induction for polarization, additionally the absence of shootin1 from the nascent axon's siblings by competition prevents the formation of surplus axons. Existing as seven alternatively spliced isoforms, the shootin1 gene is conserved in chimpanzee, dog, mouse, rat, chicken and zebrafish, and maps to human chromosome 10q25.3.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Shootin/KIAA1598
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat