To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Early B-cell Factor 2
- Antigen Symbol:EBF2
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Antigen Synonyms:COE2|OE-3|O/E-3|EBF-2
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10266-326
- Supplier no.:BS-11740R-HRP
Specifications
About this item
Early B-cell factor 2 is a 575 amino acid protein belonging to the COE family of proteins, whose members are all helix-loop-helix transcription factors. EBF2 is a transcription factor which, in synergy with the Wnt-responsive LEF1/CTNNB1 pathway, activates the decoy receptor for RANKL, OPG, in osteoblasts. OPG, in turn, regulates osteoclast differentiation. Lack of EBF2 has been found to cause a small defect in the terminal differentiation of osteoblasts, along with reduced bone mass and an increase in osteoclasts. Localized to the nucleus, EBF2 forms a homodimer or a heterodimer with a related family member.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: EBF2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat