To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:SET Domain, Bifurcated 1
- Antigen Symbol:SETDB1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Antigen Synonyms:Similar to DNA topoisomerase II.|H3 lysine 9 specic 4|ESET|Histone lysine N methyltransferase|KMT1E|SETB1_HUMAN|SETB|MGC90670|ERG-associated protein with SET domain|Histone-lysine N-methyltransferase SETDB1|Set domain protein|KIAA0067|Lysine N-methyltransferase 1E|Histone H3-K9 methyltransferase 4|Histone H3 K9 methyltransferase 4|SET domain burcated 1|KIAA00067|H3 K9 HMTase 4|mKIAA0067|KG1T|1|SETDB1|burcated|H3-K9-HMTase 4|H3-K9-HMTase4
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10261-426
- Supplier no.:BS-11670R-FITC
Specifications
About this item
ESET is a nuclear protein belonging to the histone-lysine methyltransferase family and to the Suvar3-9 subfamily. It is a highly conserved protein of 150 amino acids that has been implicated in chromatin structure modulation. ESET is excluded from cell nucleoli and areas of condensed chromatin and can associate with the nonpericentromeric regions of chromatin. The gene encoding for this protein, SETDB1, maps to chromosome 1q21. ESET is a histone methyltransferase, methylating Lys-9 of histone H3 and mutations within the SETDB1 gene abolishes its methyltransferase activity. This methylation acts as a tag for epigenetic transcriptional repression by rounding up HP1 proteins to methylated histones. ESET is widely expressed with highest levels found in testis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SETDB1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat