To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Cyclin-dependent Kinase 17
- Antigen Symbol:CDK17
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Antigen Synonyms:PCTAIRE-2|PCTK2|PCTAIRE-mot protein kinase 2|Protein kinase cdc2-related PCTAIRE-2|6430598J10Rik|CDK17_HUMAN.|MGC25109|Serine/threonine-protein kinase PCTAIRE-2|EC 2.7.1.37
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10262-518
- Supplier no.:BS-11574R-CY5
Specifications
About this item
Cell cycle progression is controlled in part by a family of cyclin proteins and cyclin dependent kinases (Cdks). Cdk proteins work in concert with cyclins to phosphorylate key substrates involved in cell cycle progression. Another family of proteins, Cdk inhibitors, also play a role in regulating the cell cycle by binding to cyclin-Cdk complexes and modulating their activity. Members of the Cdk family include Cdk2–Cdk8, PCTAIRE-1–3, PITALRE and PITSLRE. PCTAIRE-1, PCTAIRE-2 and PCTAIRE-3 comprise a subfamily of cdc2-related serine/threonine kinases. PCTAIRE-1, which is expressed primarily in mammalian brain, interacts with a variety of proteins and is thought to be part of a multiple signal transduction cascade. PCTAIRE-2, also expressed in brain, may be important in terminally differentiated neurons.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CDK17
Clonality: Polyclonal
Clone: 0
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: