To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Glial Cell Line Derived Neurotrophic Factor Family Receptor alpha 3
- Antigen Symbol:GFRA3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Antigen Synonyms:GFR alpha 3|Y15110|GDNF receptor alpha-3|GFRalpha3|GDNF family receptor alpha-3
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10258-096
- Supplier no.:BS-11501R-A488
Glial cell line-derived neurotrophic factor (GDNF) and the related neurotrophic factor neurturin (NTN) are potent survival factors for central and peripheral neurons. GDNF is a glycosylated, disulfide-bonded homodimer that is distantly related to the TGF Beta superfamily of growth factors. Three receptors for these factors, GFR Alpha-1 (also designated GDNFR-Alpha, RETL1 or TrnR-1), GFR Alpha-2 (also designated GDNFR-Beta, RETL2, NTNR-Alpha or TrnR-2) and GFR Alpha-3 have been identified. The receptors do not contain transmembrane domains and are attached to the cell membrane by glycosyl-phosphoinositol linkage. Both GFR Alpha-1 and GFR Alpha-2 have been shown to mediate the GDNF-dependent and NTN-dependent phosphorylation and activation of the tyrosine kinase Ret. GFR Alpha-3 is expressed only during development.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GFRA3/GDNF Receptor alpha 3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat