To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:Ephexin-1/EPHEXIN
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10255-988
- Supplier no.:BS-11458R-CY3
Specifications
About this item
EPHEXIN is a 710 amino acid protein that localizes to both the membrane and the cytoplasm and contains one SH3 domain, one PH domain and one DH domain. Expressed at high levels in brain and present at lower levels in lung tissue, EPHEXIN interacts with EphA4 and functions as a guanine nucleotide exchange factor (GEF) that is capable of activating Rho A, Rac 1 and Cdc42 and is thought to play a role in axon guidance and growth cone collapse. EPHEXIN is subject to Src-dependent phosphorylation, an event that increases the GEF activity of EPHEXIN toward Rho A. Human EPHEXIN, which exists as multiple alternatively spliced isoforms, shares a high degree of sequence homology with its mouse counterpart, suggesting a conserved role between species.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Ephexin-1/EPHEXIN
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat