About this item
Designed for use with WAVE Bioreactor systems, the Cellbag bioreactors are presterilized, single-use bags for noninvasive cell cultivation in research, development, and commercial manufacturing operations.
- Presterile, single-use chambers
- Fnon-invasive mixing of fluids using a WAVE Bioreactor instrument/system
- Provides high mechanical strength and bio-inert fluid contact
- Fluid contact layer is a USP Class VI ethylene vinyl acetate/low density polyethylene copolymer
- Outer noncontact layer is made of low density polyethylene
Cellbag bioreactor components are similar to those used for biological storage bags and meet USP Class VI specifications for plastics. Validation data and Cellbag DMF are available to demonstrate biocompatibility.
Each Cellbag bioreactor has an air inlet and outlet filter, a needleless sampling port, an Oxywell2 dissolved oxygen probe insertion port, and a fill/harvest port. In addition, each Cellbag bioreactor can be customized with optional fittings such as ReadyMate aseptic connectors, dip tubes, an optical pH sensor embedded in the bag, screw cap ports, temperature ports, perfusion filters, and special tubing ports.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 101725-184
- 101725-192
- 101725-178
- 101725-180
- Port:
- Needleless self-sealing syringe port
- Needleless self-sealing syringe port
- Needleless self-sealing syringe port
- Needleless self-sealing syringe port
- Operating Pressure:
- 0.1 bar (1.5 psi)
- 0.1 bar (1.5 psi)
- 0.1 bar (1.5 psi)
- 0.1 bar (1.5 psi)
- Operating Temperature:
- 50 °C
- 50 °C
- 50 °C
- 50 °C
- Thickness:
- >67 N/cm
- >67 N/cm
- >67 N/cm
- >67 N/cm