Order Entry
United States
ContactUsLinkComponent
Survivor Emergency Flashlight, Emergency Medical International
Survivor Emergency Flashlight, Emergency Medical International
Catalog # 10755-864
undefined
Survivor Emergency Flashlight, Emergency Medical International
Catalog # 10755-864
Supplier Number:  256

Specifications

  • Description:
    Emergency Wind-up Flaslight, Black
  • Environmentally Preferable:
  • Dimensions:
    61/2L x 2W x 2D"
  • Weight:
    10 oz.
  • Cat. No.:
    10755-864
  • Supplier no.:
    256

Specifications

About this item

This wind-up flashlight is a hand held battery free flashlight

It combines radio, alarm, led flashlight and cell phone charger in one. It does not need any external power source. With the Survivor you can generate light, charge mobile phones and listen to the radio anywhere, anytime with the built in wind-up handle. It can generate up to one hour of power.