ContactUsLinkComponent
Gram Stain Chemical Set
Gram Stain Chemical Set
Catalog # 470301-180
Supplier:  Ward's Science
undefined
Gram Stain Chemical Set
Catalog # 470301-180
Supplier:  Ward's Science
Supplier Number:  GG0221-PK7
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Gram Stain Chemical Set
  • Size:
    Pack of 7
  • Environmentally Preferable:
  • Chemical/physical specifications:
    30 ml Each
  • Cat. No.:
    470301-180
  • Grade:
    Laboratory
  • Supplier no.:
    GG0221-PK7
Specifications
About this item
Use these stains in the first step to identifying your unknown bacteria.
Delivery: Set includes 1 bottle of crystal violet, 1 bottle of Iodine, 1 bottle of safranin O, and 4 bottles of ethanol.
Caution: Please Note: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.