Order Entry
United States
ContactUsLinkComponent
Anti-BCL2A1 Rabbit Polyclonal Antibody
Anti-BCL2A1 Rabbit Polyclonal Antibody
Catalog # 10800-346
undefined
Anti-BCL2A1 Rabbit Polyclonal Antibody
Catalog # 10800-346
Supplier Number:  600-401-Z60
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    BCL2-Related Protein A1
  • Antigen Symbol:
    BCL2A1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Environmentally Preferable:
  • Epitope:
    N-Terminal
  • Format:
    IgG
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    597
  • Antigen Synonyms:
    Bcl2-L-5|Bcl-2-related protein A1|GRS|BCL2L5|Bcl-2-like protein 5|ACC-1|BFL1|ACC-2|HBPA1|Bfl-1
  • Concentration:
    1 mg/mL by UV absorbance at 280 nm
  • Shipping Temperature:
    Dry Ice
  • Immunogen:
    Anti-Bfl-1 antibody was prepared from whole rabbit serum produced by repeated immunizations with a 14 amino acid synthetic peptide from near the N-terminus of human Bfl-1.
  • Purification:
    Anti-Bfl-1 Antibody was affinity purified from monospecific antiserum by immunoaffinity chromatography. Cross reactivity with Bfl-1 from other sources has not been determined.
  • Cat. No.:
    10800-346
  • Supplier no.:
    600-401-Z60

Specifications

About this item

Apoptosis plays a major role in normal organism development, tissue homeostasis, and removal of damaged cells and is caused by caspase activation. Proteins that comprise the Bcl-2 family appear to control the activation of these enzymes. One such member is multi-domain antiapoptotic protein Bfl-1, which is overexpressed in stomach and other cancers. Bfl-1 can interact with Bax and suppress apoptosis by inhibiting the release of cytochrome c and caspase-3 activation. It is upregulated in cisplatin-resistant human bladder tumors, suggesting that its expression may be important for cisplatin resistance and inhibition of apoptosis in cancer cells. At least two isoforms of Bfl-1 are known to exist.

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: 10 µg/mL; Immunofluorescence Microscopy: 20 µg/mL; Western Blot: 1 - 2 µg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: BCL2A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat