Order Entry
United States
ContactUsLinkComponent
Anti-CLDN1 Rabbit Polyclonal Antibody
Anti-CLDN1 Rabbit Polyclonal Antibody
Catalog # 10800-832
undefined
Anti-CLDN1 Rabbit Polyclonal Antibody
Catalog # 10800-832
Supplier Number:  600-401-AL5
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    claudin 1
  • Antigen Symbol:
    CLDN1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Environmentally Preferable:
  • Epitope:
    C-Terminal
  • Format:
    IgG
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    9076
  • Antigen Synonyms:
    ILVASC|SEMP1|CLD1
  • Concentration:
    1 mg/mL by UV absorbance at 280 nm
  • Shipping Temperature:
    Dry Ice
  • Immunogen:
    Anti-CLDN1 antibody was prepared from whole rabbit serum produced by repeated immunizations with a 20 amino acid synthetic peptide near the C-terminus of human CLDN1.
  • Purification:
    Anti-CLDN1 Antibody was affinity purified from monospecific antiserum by immunoaffinity chromatography. Cross reactivity with CLDN1 from other sources has not been determined.
  • Cat. No.:
    10800-832
  • Supplier no.:
    600-401-AL5

Specifications

About this item

Claudin1 (CLDN1), a member of the claudin family, is an integral membrane protein and a component of tight junction strands. Tight junctions are specialized regions of cell to cell contact consisting of networking strands that act as a molecular gasket for preventing the leakage of ions, water, etc., between cells. They are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. Different tissues exhibit different Claudin composition and CLDN1 expression is often cell type and tissue dependent. Loss of function mutations result in neonatal ichthyosis-sclerosing cholangitis syndrome. CLDN1 and CLDN2 were found to be overexpressed in colonal cancer tissues and may be useful as tumor markers and targets for the treatment of colorectal cancer. Characterization of Claudins expression in human tumors can be an additional diagnostic tool. Recent studies show that CLDN1 has gastric tumor suppressive activity and is a direct transcriptional target of RUNX3. Along with SCARB1, LDL-R, and the tetraspanin superfamily member CD81, CLDN1 has been reported to be an entry factor for the Hepatitis C virus.

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: 5 µg/mL; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 1-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CLDN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: