Order Entry
United States
ContactUsLinkComponent
Anti-CALD1 Mouse Monoclonal Antibody [clone: CD-7]
Anti-CALD1 Mouse Monoclonal Antibody [clone: CD-7]
Catalog # 10800-500
Anti-CALD1 Mouse Monoclonal Antibody [clone: CD-7]
Catalog # 10800-500
Supplier Number:  200-301-P24
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Caldesmon 1
  • Antigen Symbol:
    CALD1
  • Clonality:
    Monoclonal
  • Clone:
    CD-7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Rabbit,
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Environmentally Preferable:
  • Format:
    IgG1
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    800
  • Antigen Synonyms:
    HCAD|caldesmon 1|NAG22|L-CAD|CDM|H-CAD|LCAD
  • Concentration:
    1.0 mg/mL by UV absorbance at 280 nm
  • Shipping Temperature:
    Dry Ice
  • Immunogen:
    Caldesmon (smooth) antibody was produced in mice by repeated immunizations with human uterus smooth muscle extract.
  • Purification:
    Anti-Caldesmon antibody was purified from mouse ascites by protein A chromatography.  This product reacts with human, mouse, and rabbit. Cross reactivity with Caldesmon from other sources has not been determined.
  • Cat. No.:
    10800-500
  • Supplier no.:
    200-301-P24

Specifications

About this item

Caldesmon is a potential actomyosin regulatory protein found in smooth muscle and nonmuscle cells. The predicted smooth-muscle polypeptide is 793 amino acids long. The high molecular weight caldesmon (h-CaD) is predominantly expressed in smooth muscles, whereas the low molecular weight caldesmon (l-CaD) is widely distributed in nonmuscle tissues and cells. Hayashi et al. (1992) demonstrated that the human CDM gene is composed of 14 exons. This antibody is suitable for researchers interested in neurodegenerative diseases, developmental signaling, GPCR signaling, and MAK Kinase Signaling.

Recommended Dilutions: ELISA: 1:20,000; Immunohistochemsitry: 1:100-1:500; Western Blot: 1-2ug/ml; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: CALD1
Clonality: Monoclonal
Clone: CD-7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rabbit