About this item
Ghost Dye Red 710 is an amine reactive viability dye, that can be used to discriminate viable from non-viable mammalian cells in flow cytometry applications.
This dye irreversibly binds free amines available on the cell surface, as well as intracellular free amines exposed in cells with compromised cell membranes.
Necrotic cells with compromised membranes will react with significantly more Ghost Dye Red 710 dye than viable cells in the same sample and therefore exhibit much greater fluorescence intensity, allowing exclusion of these cells from analysis.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Host:
- Environmentally Preferable: