Baseline® Tactile Monofilaments Sensory Evaluators
Catalog # 470119-634
Supplier: American 3B Scientific
Specifications
- Description:Tactile Monofilaments Sensory Evaluators Set, 20 Pieces
- Environmentally Preferable:
- Cat. No.:470119-634
- Supplier no.:W54065
Specifications
About this item
Quantify Sensation Levels.
- Units measure cutaneous sensory perception thresholds
- Each monofilament represents a unique amount of force
- Twenty monofilaments included
- Protective case included
These evaluators help test sensation levels and obtain objective data for accurate reporting on the status of diminishing or retuning sensibility. They measure cutaneous sensory perception thresholds. Each monofilament represents a unique amount of force ranging from 1.65 to 6.65 in size and .008 to 300 grams of force. The full set stores in a protective case.