To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Fission 1
- Antigen Symbol:FIS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:Liquid
- Gene ID:51024
- Antigen Synonyms:H_NH0132A01.6|TTC11|hFis1|CGI-135
- Storage Buffer:PBS containing 20% glycerol, 0.2% BSA, and 0.01% thimerosal.
- Storage Temperature:-20 °C
- Shipping Temperature:4°C
- Immunogen:Synthetic peptide surrounding amino acid 143 of human Fis1
- Purification:Affinity purified
- Cat. No.:10004-138
- Supplier no.:3491-100
hFis1 is a novel molecule that was discovered as part of the mammalian fission machinery. Fis 1 has been sμggested to play a role in regulation of the fission processes that might be involved in apoptotic mechanisms.
Recommended Dilutions: Western Blot: 1:100-500; Immunoprecipitation: 1:50-200; Immunohistochemistry: 1:100; Iimmunofluorescence: 1:20-100
Type: Primary
Antigen: Fis1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat