Ethyl-3-hydroxy-3-phenylpropionate 95%
About this item
Purity: 95% CAS No.: 5764-85-2 Molecular Formula: C11H14O3 Formula Weight: 194.23 Physical Form: Solid Density: 1.088 Refractive Index: 1.5080 Beilstein Reference: 2211602 MDL No.: MFCD00086891
Synonyms: Ethyl 3-hydroxy-3-phenylpropanoate
- CAS Number:5764-85-2
- Formula:C₁₁H₁₄O₃
- MDL Number:MFCD00086891
- Molecular Weight:194.23 g/mol
- Density:1.088
- Storage Temperature:Ambient
- EINECS:227-288-2
0 Options Available Below
Product Details & Documents
Purity: 95% CAS No.: 5764-85-2 Molecular Formula: C11H14O3 Formula Weight: 194.23 Physical Form: Solid Density: 1.088 Refractive Index: 1.5080 Beilstein Reference: 2211602 MDL No.: MFCD00086891
Synonyms: Ethyl 3-hydroxy-3-phenylpropanoate
- CAS Number:5764-85-2
- Formula:C₁₁H₁₄O₃
- MDL Number:MFCD00086891
- Molecular Weight:194.23 g/mol
- Density:1.088
- Storage Temperature:Ambient
- EINECS:227-288-2
Documents
Specifications for Product Family
Specification Test Results
- Assay Minimum:95
Product Family Options
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



