ContactUsLinkComponent
Manual Injection Valve, Cytiva
Manual Injection Valve, Cytiva
Catalog # 10498-050
Supplier:  Cytiva
Product available on GSA Advantage®
undefined
Manual Injection Valve, Cytiva
Catalog # 10498-050
Supplier:  Cytiva
Supplier Number:  29-0239-58
Product available on GSA Advantage®
Specifications
  • Description:
    Manual Injection Valve
  • Environmentally Preferable:
  • Cat. No.:
    10498-050
  • Supplier no.:
    29-0239-58
Specifications
About this item
Manual injection valve is a 6-port rotary valve that is manually operated to transfer the preloaded sample onto the column.