Digivac TracVac Progressive Display Vacuum Gauge, Ace Glass Incorporated
Catalog # 10767-866
Supplier: Ace Glass
Small Business Enterprise
Specifications
- Description:Digivac TracVacMeter, 100-240V AC 50/60Hz
- Environmentally Preferable:
- Cat. No.:10767-866
- Supplier no.:14301-01
Specifications
About this item
A bar graph style vacuum meter that visually illustrates vacuum pressure rate changes to enable the quick determination of increasing or decreasing vacuum
The vacuum interface is 1/8" NPT or 1/4" male flare. The meter includes a 10 foot sensor cord.
The range is 1-760,000 micron (milliTorr).
Certifications: CE rating.