To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antigen Name:Neurotrophic Tyrosine Kinase Receptor, Type 2
- Antigen Symbol:NTRK2
- Host:Multiple
- Isotype:
- Reactivity:Human
- Size:1 Set
- Environmentally Preferable:
- Gene ID:4915
- Antigen Synonyms:GP95-TrkB|GP145-TrkB/GP95-TrkB|GP95 TrkB|BDNF/NT-3 growth factors receptor|EC 2.7.10.1|Trk-B|BDNF/NT 3 growth factors receptor|GP145-TrkB|Tkrb|AI848316|Neurotrophin receptor tyrosine kinase type 2|NTRK2|TRKB1|NTRK 2|GP145 TrkB|included|RATTRKB1|Obesity|C030027L06Rik|Trk B|hyperphagia|and developmental delay|TrkB tyrosine kinase|TRKB|neurotrophic tyrosine kinase receptor type 2|BDNF tropomyosine receptor kinase B|Brain derived neurotrophic factor receptor|Tropomyosin related kinase
- Cat. No.:10540-738
- Supplier no.:H00004915-AP52
This Set comes with matched Antibody Pair to detect and quantify protein level of human NTRK2.
Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-NTRK2 (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-NTRK2 (50 µg);
Type:
Antigen: NTRK2
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human