To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Name:Polymerase (DNA Directed), Delta 2
- Antigen Symbol:POLD2
- Clonality:Polyclonal
- Host:Multiple
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Gene ID:5425
- Cat. No.:10541-712
- Supplier no.:H00005425-AP21
Specifications
About this item
This Set comes with matched Antibody Pair to detect and quantify protein level of human POLD2.
Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-POLD2 (100 µg); 2. Detection Antibody: Mouse purified Polyclonal Anti-POLD2 (20 µg);
Type:
Antigen: POLD2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human