To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antigen Name:coiled-coil Domain Containing 6
- Antigen Symbol:CCDC6
- Host:Multiple
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Gene ID:8030
- Antigen Synonyms:D10S170|H4|TPC|TST1|PTC
- Cat. No.:10536-614
- Supplier no.:H00008030-AP11
This Set comes with matched Antibody Pair to detect and quantify protein level of human CCDC6.
Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-CCDC6 (100 µg); 2. Detection Antibody: Mouse Monoclonal Anti-CCDC6, IgG1 kappa (20 µg);
Type:
Antigen: CCDC6
Clonality: 0
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype: IgG1
Reactivity: Human