To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Name:Fibroblast Activation Protein Alpha
- Antigen Symbol:FAP
- Host:Multiple
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Gene ID:2191
- Antigen Synonyms:SIMP|DPPIV|FAPA
- Cat. No.:10538-232
- Supplier no.:H00002191-AP51
Specifications
About this item
This Set comes with matched Antibody Pair to detect and quantify protein level of human FAP.
Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-FAP (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-FAP (50 µg);