To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Name:Calcium/calmodulin-dependent Protein Kinase 2 alpha
- Antigen Symbol:CAMK2A
- Host:Multiple
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Gene ID:815
- Antigen Synonyms:CaM kinase II alpha subunit|CaMK-II alpha subunit|CaM-kinase II alpha chain|CAMKA|PKCCD|PK2CDD|Calcium/calmodulin-dependent protein kinase type II alpha chain
- Modification:phospho-Thr286
- Cat. No.:10540-814
- Supplier no.:DP0227
Specifications
About this item
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the CAMK2A protein, and the other against the specific T286 phosphorylated site of CAMK2A for use in Proximity Ligation Assay
Antibody pair set content: 1. Phospho-CAMK2A T286 Rabbit Polyclonal Antibody (20 µl) In PBS, 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. CAMK2A Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Type:
Antigen: CAMK2A
Clonality: 0
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype: IgG1
Reactivity: