Anti-SMT3 Mouse Monoclonal Antibody [clone: 4F2.F5.G2]
Catalog # RL200-301-428
Supplier: Rockland Immunochemical
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Small Ubiquitin-related Modifier 3
- Antigen Symbol:SMT3
- Clonality:Monoclonal
- Clone:4F2.F5.G2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Yeast
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:852122
- Antigen Synonyms:SUMO-2|ubiquitin-like protein SMT3A|SMT3 homolog 1|ubiquitin-like protein SMT3B|SMT3H1|SMT3 suppressor of mif two 3 homolog 1|Smt3B|SUMO-3|SMT3A|small ubiquitin-related modifier 3
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage Temperature:–20 °C
- Concentration:1.9 mg/ml by UV absorbance at 280 nm
- Shipping Temperature:Dry Ice
- Immunogen:This antibody was produced in mice by repeated immunizations with full-length recombinant yeast SUMO Protein.
- Purification:Protein A purified
- Cat. No.:RL200-301-428
- Supplier no.:200-301-428
Specifications
About this item
Type: Primary
Antigen: SMT3
Clonality: Monoclonal
Clone: 4F2.F5.G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Yeast