Brilliant Violet 421™ anti-human CD366 (Tim-3) [F38-2E2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Rhesus; Apps: FC; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD366 (Tim-3)
Clonality: Monoclonal
Clone: F38-2E2
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Rhesus
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 345007-BL
- 345008-BL
- Antigen Symbol:
- CD366 (Tim-3)
- CD366 (Tim-3)
- Antigen Synonyms:
- Th1|hepatitis virus cellular receptor 2|CD366 Havcr2|Timd3|Tim 3|Immune checkpoints|T cell immunoglobulin and mucin domain containing protein 3|Tim3|HAVCR2
- Th1|hepatitis virus cellular receptor 2|CD366 Havcr2|Timd3|Tim 3|Immune checkpoints|T cell immunoglobulin and mucin domain containing protein 3|Tim3|HAVCR2
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Brilliant Violet® 421
- Brilliant Violet® 421
- Clone:
- F38-2E2
- F38-2E2
- Reactivity:
- Rhesus monkey, Human
- Rhesus monkey, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 84868
- 84868
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow Cytometry:
- Yes
- Immunogen:
- Human Tim-3 fusion protein
- Human Tim-3 fusion protein
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Purification:
- The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 421™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 421™ and unconjugated antibody.
- The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 421™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 421™ and unconjugated antibody.
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT