Order Entry
United States
ContactUsLinkComponent
Anti-IgEb Rat Monoclonal Antibody (Biotin) [clone: JKS-6]
Anti-IgEb Rat Monoclonal Antibody (Biotin) [clone: JKS-6]
  408704-BL
 :  Biolegend
Anti-IgEb Rat Monoclonal Antibody (Biotin) [clone: JKS-6]
  408704-BL
 :  Biolegend
 :  408704
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Symbol:
    IgEb
  • Clonality:
    Monoclonal
  • Clone:
    JKS-6
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Gel Shift:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Mouse
  • Size:
    500 μg
  • Environmentally Preferable:
  • Form:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Gene ID:
    16020
  • Antigen Synonyms:
    IgE
  • Storage Temperature:
    The antibody solution should be stored undiluted between 2 °C and 8 °C.
  • Concentration:
    0.50 mg/mL
  • Shipping Temperature:
    Ambient RT
  • Purification:
    The antibody was purified by affinity chromatography, and conjugated with biotin under optimal conditions. The solution is free of unconjugated biotin.
  • Cat. No.:
    408704-BL
  • Supplier no.:
    408704

 

 

Biotin anti-mouse IgEb [JKS-6]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ELISA, FC; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA. For ELISA detection, the suggested use of this reagent is 1-2 μg per ml. It's recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IgEb
Clonality: Monoclonal
Clone: JKS-6
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse