Order Entry
United States
ContactUsLinkComponent
Anti-PRC1 Mouse Monoclonal Antibody [clone: 6G2]
Anti-PRC1 Mouse Monoclonal Antibody [clone: 6G2]
Supplier:  Biolegend
Certificates
Anti-PRC1 Mouse Monoclonal Antibody [clone: 6G2]
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • 629002-BL
  • 629001-BL
  • Antigen Symbol:
  • PRC1
  • PRC1
  • Antigen Synonyms:
  • Mitotic spindle-associated CDK substrate|Protein regulating cytokinesis 1|Protein regulator of cytokinesis 1
  • Mitotic spindle-associated CDK substrate|Protein regulating cytokinesis 1|Protein regulator of cytokinesis 1
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 6G2
  • 6G2
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 9055
  • 9055
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • Western Blot:
  • Yes
  • Yes
  • Immunogen:
  • Amino Acid: 1-150
  • Amino Acid: 1-150
  • Form:
  • This antibody is provided in phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide at 0.5 mg/ml.
  • This antibody is provided in phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide at 0.5 mg/ml.
  • Purification:
  • The antibody was purified by affinity chromatography.
  • The antibody was purified by affinity chromatography.
  • Concentration:
  • 0.50 mg/mL
  • 0.50 mg/mL
  • Storage Temperature:
  • The antibody solution should be stored undiluted between 2 °C and 8 °C.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C.
  • Shipping Temperature:
  • Ambient RT
  • Ambient RT

Specifications

Related Information

Frequently Bought Together