About this item
Alexa Fluor® 647 anti-human CD163 [RM3/1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, IF; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD163
Clonality: Monoclonal
Clone: RM3/1
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 326507-BL
- 326508-BL
- Antigen Symbol:
- CD163
- CD163
- Antigen Synonyms:
- Hemoglobin/Haptoglobin Complex Receptor|M130|ED2(rat)|Macrophage marker|GHI/61|RM3/1|p155|macrophage-associated antigen
- Hemoglobin/Haptoglobin Complex Receptor|M130|ED2(rat)|Macrophage marker|GHI/61|RM3/1|p155|macrophage-associated antigen
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Alexa Fluor® 647
- Alexa Fluor® 647
- Clone:
- RM3/1
- RM3/1
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 9332
- 9332
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- Human monocytes
- Human monocytes
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with Alexa Fluor® 647 under optimal conditions.
- The antibody was purified by affinity chromatography, and conjugated with Alexa Fluor® 647 under optimal conditions.
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT