Order Entry
United States
ContactUsLinkComponent
Anti-HaloTag Rabbit Polyclonal Antibody
Anti-HaloTag Rabbit Polyclonal Antibody
Catalog # PAG9281
undefined
Anti-HaloTag Rabbit Polyclonal Antibody
Catalog # PAG9281
Supplier Number:  G9281

Specifications

  • Antibody Type:
    Tag
  • Antigen Name:
    HaloTag
  • Antigen Symbol:
    HaloTag
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
  • Size:
    200 μg
  • Environmentally Preferable:
  • Storage Temperature:
    –30 °C to –10 °C
  • Cat. No.:
    PAG9281
  • Supplier no.:
    G9281

Specifications

About this item

Anti-HaloTag pAb is a purified rabbit polyclonal antibody raised against the HaloTag protein

  • Polyclonal Rabbit Antibody for Detecting HaloTag(R) Fusion Proteins
  • Specific for HaloTag(R) protein
  • Exhibits low cross-reactivity with E. coli and mammalian cell extracts
  • Supplied at a concentration of 1mg/ml

The Anti-HaloTag pAb is a purified rabbit polyclonal antibody raised against the HaloTag protein. The antibody is purified using Protein G affinity resin. The antibody detects HaloTag fusion proteins in Western blot hybridization and immunocytochemistry applications with high sensitivity and specificity. The HaloTag protein is not endogenous to mammalian, plant and E. coli cells. E. coli and mammalian cell extracts demonstrate low cross-reactivity with the Anti-HaloTag pAb.

Type:
Antigen: HaloTag
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: