Modified Koppers Vacuum Viscometer, Cannon Instrument
Supplier: Cannon
Certificates
About this item
The Modified Koppers Vacuum Viscometer measures the viscosity of highly viscous materials
Viscometer is calibrated. Minimum sample volume 2 mL. Minimum bath depth 7".
Frequently Bought Together