Order Entry
United States
ContactUsLinkComponent
Catalase Activity Colorimetric/Fluorometric Assay Kit, BioVision
Catalase Activity Colorimetric/Fluorometric Assay Kit, BioVision
Catalog # 10187-026
Supplier:  Avantor
CAS Number:  
undefined
Catalase Activity Colorimetric/Fluorometric Assay Kit, BioVision
Catalog # 10187-026
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    100 assays
  • Environmentally Preferable:
  • Species Reactivity:
    Mammalian samples
  • Shelf Life:
    12 Months
  • Sample Size:
    Cell and tissue culture supernatants, urine, plasma and serum, as well as many other biological fluids, growth medium, food, etc.
  • Storage Temperature:
    -20°C
  • Cat. No.:
    10187-026
  • Supplier no.:
    K773-100

Specifications

About this item

Highly Sensitive Assay, HTS

Detection method- Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Species reactivity- Mammalian
Applications- The kit detects high pico-unit of catalase in samples.

Catalase (EC 1.11.1.6) is a ubiquitous antioxidant enzyme that is present in nearly all living organisms. It functions to catalyze the decomposition of hydrogen peroxide (H₂O₂) to water and oxygen. BioVision’s Catalase Assay Kit provides a highly sensitive, simple, direct and HTS-ready assay for measuring Catalase activity in biological samples. In the assay, catalase first reacts with H₂O₂ to produce water and oxygen, the unconverted H₂O₂ reacts with OxiRed™ probe to produce a product, which can be measured at 570 nm (Colorimetric method) or at Ex/Em=535/587nm (fluorometric method). Catalase activity is reversely proportional to the signal. The kit detects high pico-unit of catalase in samples.