Order Entry
United States
ContactUsLinkComponent
Listeria Enrichment Broth (LEB) acc. to FDA/IDF-FIL, MilliporeSigma
Listeria Enrichment Broth (LEB) acc. to FDA/IDF-FIL, MilliporeSigma
Catalog # EM1.10549.5000
Supplier:  MilliporeSigma
CAS Number:  
Listeria Enrichment Broth (LEB) acc. to FDA/IDF-FIL, MilliporeSigma
Catalog # EM1.10549.5000
Supplier:  MilliporeSigma
Supplier Number:  1.10549.5000
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Plastic drum
  • Description:
    Listeria Enrichment Broth
  • Size:
    5 kg
  • Environmentally Preferable:
  • Incubation:
    24 hrs; 30 °C; aerobically
  • pH:
    7.1 to 7.5 at 25°C
  • Cat. No.:
    EM1.10549.5000
  • Supplier no.:
    1.10549.5000

Specifications

About this item

In North America 1.10549.5000 replaces and is identical to 1.10549.5007

  • 10 to 100 inoculum on reference medium Listeria monocytogenes ATCC 13932 (WDCM 00021)
  • 10 to 100 inoculum on reference medium Listeria monocytogenes ATCC 19114 (WDCM 00110)
  • 10 to 100 inoculum on reference medium Listeria monocytogenes ATCC 35152 (WDCM 00109)
  • 10 to 100 inoculum on reference medium Listeria innocua ATCC 33090 (WDCM 00017)
  • Greater than or equal to 1E+04 CFU/mL colony count in Listeria monocytogenes ATCC 13932 (WDCM 00021)
  • Greater than or equal to 1E+04 CFU/mL colony count in Listeria monocytogenes ATCC 19114 (WDCM 00110)
  • Greater than or equal to 1E+04 CFU/mL colony count in Listeria monocytogenes ATCC 35152 (WDCM 00109)
  • Greater than or equal to 1E+04 CFU/mL colony count in Listeria innocua ATCC 33090 (WDCM 00017)
  • Less than or equal to 1000 CFU/mL colony count in Staphylococcus aureus ATCC 6538 (WDCM 00032)