Order Entry
United States
ContactUsLinkComponent
Human Recombinant CD93 (from HEK293 Cells)
Human Recombinant CD93 (from HEK293 Cells)
Catalog # 10797-082
Supplier:  Avantor
CAS Number:  
undefined
Human Recombinant CD93 (from HEK293 Cells)
Catalog # 10797-082
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Environmentally Preferable:
  • Tag sequence:
    His Tag
  • Protein Synonyms:
    C1qRP|CD93|C1QR1|CDw93|MXRA4|ECSM3|C1qR(P)|C1qR
  • Protein/Peptide Name:
    CD93
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    59 kDa
  • Formulation:
    Lyophilized, PBS, pH7.4
  • Tested Applications:
    Western Blot
  • Cat. No.:
    10797-082
  • Supplier no.:
    96-069

Specifications

About this item

CD antigen CD93 is also known as Complement component C1q receptor (C1QR1). which belongs to the Group XIV C-Type lectin family, a group containing two other members, endosialin (CD248) and thrombomodulin. CD93 contains one C-type lectin domain and five EGF-like domains. CD93 is highly expressed in endothelial cells, platelets, cells of myeloid origin, such as monocytes and neutrophils. CD93 is receptor for C1q, mannose-binding lectin (MBL2) and pulmonary surfactant protein A (SPA). CD93 may mediate the enhancement of phagocytosis in monocytes and macrophages upon interaction with soluble defense collagens and may play a role in intercellular adhesion.