Order Entry
United States
ContactUsLinkComponent
 
Anti-FAM62A Goat Polyclonal Antibody
  89296-802
 :  Genetex
Anti-FAM62A Goat Polyclonal Antibody
  89296-802
 :  Genetex
 :  GTX88786
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Family With Sequence Similarity 62, Member A
  • Antigen Symbol:
    FAM62A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Bovine,
    Dog,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 μg
  • Environmentally Preferable:
  • Gene ID:
    23344
  • Antigen Synonyms:
    FAM62A|MBC2
  • Storage Buffer:
    Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
  • Storage Temperature:
    Aliquot and store at –20 °C. Minimize freezing and thawing.
  • Concentration:
    0.5mg/ml
  • Immunogen:
    C-QKKRTLSPEFNER
  • Purification:
    Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
  • Cat. No.:
    89296-802
  • Supplier no.:
    GTX88786

 

 

Goat polyclonal antibody to FAM62A

Approx. 150kDa band observed in Human Adipose lysates (calculated MW of 123kDa according to NP_056107.1). In transfected HEK293 transiently expressing FAM62A a band of approx. 120kDa is observed. This band is not observed in the non-transfected HEK293. Recommended concentration: 1-3µg/ml.

Type: Primary
Antigen: ESYT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human