To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Apolipoprotein B mRNA Editing Enzyme Catalytic Polypeptide-Like 3G
- Antigen Symbol:APOBPEC3G
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:60489
- Antigen Synonyms:dJ494G10.1|bK150C2.7|ARP9|ARCD|A3G|CEM-15|CEM15|MDS019|ARP-9
- Storage Buffer:Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
- Storage Temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:0.5mg/ml
- Immunogen:C-DEHSQDLSGRLR
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:89296-530
- Supplier no.:GTX88548
Specifications
About this item
Goat polyclonal antibody to APOBEC3G
Approx. 45kDa band observed in lysates of cell line Daudi (calculated MW of 46.4kDa according to NP_068594.1). Recommended concentration: 0.5-1.5µg/ml.
Type: Primary
Antigen: APOBPEC3G
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human