To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:CUB And Sushi Multiple Domains 1
- Antigen Symbol:CSMD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Bovine,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:64478
- Antigen Synonyms:PPP1R24
- Storage Buffer:Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
- Storage Temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:0.5mg/ml
- Immunogen:C-KGVREVNETVTKTP
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:89296-174
- Supplier no.:GTX88248
Specifications
About this item
Goat polyclonal antibody to CSMD1
Approx 150kDa band observed in Human Brain (Cerebellum) lysates (calculated MW of 389kDa according to NP_150094.4). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Kraus et al, J Immunol. 2006 Apr 1;176(7):4419-30. PMID: 16547280). Recommended concentration: 1-3µg/ml.
Type: Primary
Antigen: CSMD1 (CUB and Sushi multiple Domains 1)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: