To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Interferon-induced Protein With Tetratricopeptide Repeats 3
- Antigen Symbol:IFIT3
- Clonality:Monoclonal
- Clone:1G1
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Gel Shift:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:3437
- Antigen Synonyms:IFIT-3|IFIT4|CIG-49|IFI60|RIG-G|Interferon-induced protein with tetratricopeptide repeats 3|IRG2|IFIT3 |P60|ISG60|GARG-49|CIG49
- Storage Buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0.02% sodium azide.
- Storage Temperature:For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human IFIT3 (NP_001540) produced in HEK293T cell.
- Purification:Purified from mouse ascites fluids by Protein A/G affinity chromatography
- Cat. No.:89273-422
- Supplier no.:GTX84314
Specifications
About this item
Mouse monoclonal antibody [1G1] to IFIT3
Recommended Starting Dilutions:WB 1:2000 IHC 1:50 ICC/IF 1:100 Flow 1:100Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: IFIT3
Clonality: Monoclonal
Clone: 1G1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human