To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:40S ribosomal protein S27
- Antigen Symbol:RPS27
- Clonality:Monoclonal
- Clone:7E3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 μl
- Environmentally Preferable:
- Gene ID:6232
- Antigen Synonyms:metallopanstimulin 1|MPS-1|MPS1|40S ribosomal protein S27|RP11-422P24.3|ribosomal protein S27
- Storage Buffer:Ascitic fluid containing 0.03% sodium azide.
- Storage Temperature:Store at 4℃, for long term storage, store at -20℃.
- Immunogen:The immunogen corresponding to MPS1 expressed in E. Coli.
- Cat. No.:89287-698
- Supplier no.:GTX83377
Specifications
About this item
Mouse monoclonal antibody [7E3] to MPS1
Recommended Starting Dilutions:ELISA: 1/10000.ICC: 1/200 - 1/1000.Not yet tested in other applications.Determining optimal working dilutions by titration test.
Type: Primary
Antigen: RPS27
Clonality: Monoclonal
Clone: 7000
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human