To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Translocase of Outer Mitochondrial Membrane 40
- Antigen Symbol:TOMM40
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:10452
- Antigen Synonyms:C19orf1|translocase of outer mitochondrial membrane 40|translocase of outer mitochondrial membrane 40 homolog (yeast)|PEREC1|TOMM40|TOM40|mitochondrial outer membrane protein|PER-EC1|D19S1177E
- Storage Buffer:PBS (pH 7.2) containing 30% glycerol, 0.5% BSA and 0.01% thimerosal
- Storage Temperature:The antibody solution should be gently mixed before use. Stored at -20 °C for future use.
- Concentration:0.5 mg/ml
- Purification:Antigen affinity purification
- Cat. No.:89283-690
- Supplier no.:GTX59900
Specifications
About this item
Rabbit polyclonal antibody to Tom 40
Western blotting (0.5-4 µg/ml). However, the optimal concentrations should be determined individually. Blocking peptide (Cat.# 3740BP-50) is available separately. The antibody recognizes TOM40 in samples of human, mouse and rat origins. Reactivity to other species has not been tested.
Type: Primary
Antigen: TOMM40
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat