To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Cd44 Molecule (Indian Blood Group)
- Antigen Symbol:HCAM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:960
- Antigen Synonyms:MDU2|HUTCH-I|LHR|MC56|Pgp1|CDW44|MIC4|ECMR-III|HCELL|MDU3|IN|CSPG8
- Storage Buffer:PBS (pH 7.2) containing 50% glycerol, 1% BSA, and 0.02% thimerosal
- Storage Temperature:The antibody solution should be gently mixed before use. Stored at -20 °C for future use.
- Concentration:0.2 mg/ml
- Purification:Antigen affinity purification
- Cat. No.:89279-850
- Supplier no.:GTX59633
Specifications
About this item
Rabbit polyclonal antibody to HCAM
Western blotting (0.5-4 µg/ml), immunoprecipitation (20 µg/ml) and Immunohistochemistry (20 µg/ml). However, the optimal concentrations should be determined individually.
Type: Primary
Antigen: CD44
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat