To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Mitogen-activated Protein kinase 9
- Antigen Symbol:SAPK/JNK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:5601
- Antigen Synonyms:SAPK1a|JNK-55|JNK2A|PRKM9|p54aSAPK|SAPK|p54a|JNK2|JNK2BETA|JNK2B|JNK2ALPHA
- Storage Buffer:saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Peptide sequence around aa.181~185 (M-M-T-P-Y) derived from Human SAPK/JNK.
- Purification:Antibodies were purified by affinity-chromatography using epitope-specific peptide.
- Cat. No.:89304-980
- Supplier no.:GTX50564
Specifications
About this item
Rabbit Polyclonal antibody to JNK
Suggested starting dilutions are as follows. WB: 1:500~1:1000, IHC-P: 1:50~1:100. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: SAPK/JNK
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat