To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Small Ubiquitin-Like Modifier 1
- Antigen Symbol:SUMO1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:S. cerevisiae
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:852122
- Antigen Synonyms:SMT3|DAP1|ubiquitin-like protein SMT3C|UBL1|sentrin|SMT3H3|GAP modifying protein 1|SMT3C|ubiquitin-like protein UBL1|SMT3 homolog 3|GMP1|OFC10|OK/SW-cl.43|PIC1|ubiquitin-homology domain protein PIC1|SENP2
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 and 0.01% (w/v) Sodium Azide
- Storage Temperature:Store vial at 4 °C for short-term use. For extended storage aliquot contents and freeze at –20 °C or below. Avoid cycles of freezing and thawing.
- Immunogen:This purified antibody was prepared from rabbit serum after repeated immunizations with recombinant yeast SUMO protein.
- Purification:IgG fraction
- Cat. No.:89333-562
- Supplier no.:GTX48822
Specifications
About this item
Rabbit Polyclonal antibody to Sumo
Recommended Starting Dilutions: WB: Use at 1:500 - 1:3,000 ELISA: Use at 1:5,000 - 1:25,000
Type: Primary
Antigen: SUMO
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: S. cerevisiae