To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Eif3S6/Int6
- Antigen Symbol:Eif3S6/Int6 (eukaryotic translation initiation Factor 3, Subunit E)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Monkey,Human,Bovine,Dog,Rat,Mouse
- Western Blot:Yes
- Size:50 μg
- Environmentally Preferable:
- Gene ID:16341
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 and 0.01% (w/v) Sodium Azide
- Storage Temperature:Store vial at –20 °C prior to opening. Aliquot contents and freeze at –20 °C or below for extended storage Avoid cycles of freezing and thawing.
- Concentration:0.8 mg/ml
- Immunogen:This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a region near the C-terminus of mouse EIF3S6/Int6.
- Purification:Purified by antigen-affinity chromatography.
- Cat. No.:89317-604
- Supplier no.:GTX48712
Rabbit Polyclonal antibody to Eif3S6
Recommended Starting Dilutions: WB: Use at 1:1,000 ELISA: Use at 1:90,000
Type: Primary
Antigen: Eif3S6/Int6 (eukaryotic translation initiation Factor 3, Subunit E)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Bovine, Dog, Monkey, Rat