To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Son Of Sevenless Homolog 1
- Antigen Symbol:SOS1
- Clonality:Monoclonal
- Clone:SOS-01
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:6654
- Antigen Synonyms:GF1|GINGF|HGF|GGF1|SOS-1|NS4
- Storage Buffer:Phosphate buffered saline containing 0.09% Sodium Azide (NaN3)
- Immunogen:Synthetic peptide corresponding to amino acids THPSMHRDGPPLLENAHSS of human SOS1.
- Purification:Purified IgG prepared by affinity chromatography on Protein A
- Cat. No.:89285-612
- Supplier no.:GTX39059
Mouse monoclonal antibody [SOS-01] to SOS1
Recommended Starting Dilutions:Western Blotting: Max Dilution 1.0 ug/mlNot tested in other applications. Optimal dilution / concentration should be determined by the end user.
Type: Primary
Antigen: SOS1
Clonality: Monoclonal
Clone: SOS-1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse