Tetrasodium diphosphate decahydrate 99.0-103.0% ACS
Supplier: BeanTown Chemical
Certificates
About this item
Synonyms: Sodium pyrophosphate decahydrate, tetra-Sodium pyrophosphate decahydrate, Sodium pyrophosphate tetrabasic decahydrate, TSPP
CAS Number:13472-36-1
:
Formula:Na₄P₂O₇·10H₂O
MDL Number:MFCD00149200
Density:1.82 g/mL
Molecular Weight:446.06 g/mol
Melting Pt:990
EINECS:231-767-1
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:99.0-103.0%
- pH of 5% solution at 25°C:9.5-10.5
- Insoluble matter:≤0.01%
- Chloride (Cl):≤0.002%
- Sulfate (SO4):≤0.005%
- Nitrogen compounds (as N):≤0.001%
- Heavy metals:≤0.001%
- Iron (Fe):≤0.001%
- Arsenic (As):≤5ppm
Specification Test Results
Frequently Bought Together