ContactUsLinkComponent
Fluoromount-G® Slide Mounting Medium, Southern Biotech
Fluoromount-G® Slide Mounting Medium, Southern Biotech
Catalog # 100241-874
CAS Number:  
Fluoromount-G® Slide Mounting Medium, Southern Biotech
Catalog # 100241-874
Supplier Number:  0100-01
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Fluoromount-G® Slide Mounting Medium
  • Size:
    25 mL
  • Environmentally Preferable:
  • Cat. No.:
    100241-874
  • Supplier no.:
    0100-01
Specifications
About this item
Fluoromount-G® is suitable for mounting frozen and paraffin sections, whole mounts, and cells.
  • For mounting frozen and paraffin sections, whole mounts, and cells
  • Water soluble
  • Semi-permanent seal
Fluoromount-G® is a water soluble compound recommended for slides mounted after a staining procedure having an aqueous final step. Because Fluoromount-G® is water soluble, the coverslip may be removed by submerging the slide in a PBS solution until the coverslip is loosened. This mounting medium also provides a semi-permanent seal for storage of slide preparations.