Order Entry
United States
Orders LinkContactUsLinkComponent
Phorbol-12-myristate-13-acetate

Phorbol-12-myristate-13-acetate

5 mg
Catalog # 10187-494
Phorbol-12-myristate-13-acetate
Catalog # 10187-494
Supplier:  
CAS Number:  16561-29-8
Phorbol-12-myristate-13-acetate
Phorbol-12-myristate-13-acetate

5 mg

CAS Number:  16561-29-8
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
PHORBOL 12-MYRISTATE 13-ACETATE 25MG Catalog #76461-506

Product 76461-506 is similar to 10187-494 but manufacturer and specifications may vary.

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Synonyms:  TPA, Phorbol 12-myristate 13-acetate, 12-O-Tetradecanoylphorbol 13-acetate, 4β,9α,12β,13α,20-Pentahydroxytiglia-1,6-dien-3-one 12-tetradecanoate 13-acetate, (1aR,1bS,4aR,7aS,7bS,8R,9R,9aS)-9a-Acetoxy-4a,7b-dihydroxy-3-(hydroxymethyl)-1,1,6,8-tetramethyl-5-oxo-1a,1b,4,4a,5,7a,7b,8,9,9a-decahydro-1H-cyclopropa[3,4]benzo[1,2-e]azulen-9-yl tetradecanoate
  • CAS Number:16561-29-8
  • Formula:C₃₆H₅₆O₈
  • MDL Number:MFCD00036736
  • Molecular Weight:616.836 g/mol

Documents

Specifications

Specification Test Results

  • CAS:
    16561-29-8
  • Purity:
    99%
  • Form:
    White solid

Specifications

  • Size:
    5 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.