To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Growth Differentiation Factor 11
- Antigen Symbol:GDF11
- Clonality:Monoclonal
- Clone:3G6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Lyophilized
- Antigen Synonyms:GDF11|Bone morphogenetic protein 11|BMP11|GDF-11|Growth/differentiation factor 11 [Precursor]
- Immunogen:Partial recombinant human GDF-11 (310-407) with a GST tag.
- Cat. No.:10782-234
- Supplier no.:M-885-100
Specifications
About this item
GDF-11 is a secreted signal that acts globally to specify positional identity along the anterior/posterior axis during development. It plays critical roles in patterning both mesodermal and neural tissues and in establishing the skeletal pattern. It exists as a disulfide-linked homodimer. It belongs to the TGF-beta family.
Specificity has been confirmed by direct ELISA against the antigen.
Application Information:
This antibody is recommended for IHC, and sandwich ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: GDF-11
Clonality: Monoclonal
Clone: 3G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human